About this item
Aqueous IFD from Cytiva is a polypropylene housed in-line filter/degasser (IFD) with nylon membrane. It connects to an HPLC line and simultaneously filters and degasses during HPLC mobile phase preparation.
- Faster than traditional methods of HPLC mobile phase preparation, saving time in the laboratory
- Provides pure filtration of aqueous mobile phases without expensive degassing equipment
- Chemically resistant polypropylene construction enables reliable filtration
- Can be integrity-tested with the bubble point method
- Includes compression end fittings
- Recyclable after use
The Aqueous IFD is designed to work as an in-line filter with mobile phases containing at least 20% of the aqueous component. It has a 0.2 µm hydrophilic nylon membrane to support aqueous-based HPLC mobile phase preparation.
Whatman Aqueous IFD have a polypropylene housing sealed by a security ring, fittings to accommodate 1/16”-1/8” tubing, and an air vent on the inlet with luer lock cap to enable priming.
An in-line filter works on the principle of "bubble point" – the point of pressure at which gases will pass through a wet membrane. If pressure is maintained below the bubble point, the gas will not pass through the membrane and is trapped by the filter device.
Specifications
- Catalog No:
- 80089-524
- Área de filtración:
- 16 cm²
- Velocidad de flujo:
- 2.5 ml/min [Max], For effective gas bubble removal in HPLC
- Housing material:
- Polypropylene (PP)
Specifications
Related Information
Certificates
Search by Lot NumberCytiva CyDye Licensing Statements Cytiva General Licensing Statements