Orders LinkContactUsLinkComponent
PYREX® EZ Access™ Volumetric Flasks, Class A, Corning
   Sustainable Options Available
Supplier:   Corning
PYREX® EZ Access™ Volumetric Flasks, Class A, Corning
Supplier:  Corning
   Sustainable Options Available
Certificates
PYREX® EZ Access™ Volumetric Flasks, Class A, Corning
PYREX® EZ Access™ Volumetric Flasks, Class A, Corning
6 Options Available Below
Product Details & Documents
PYREX® glass, low actinic, with stopper.
  • Graduation line is sharp and permanent
  • Low actinic glass provides protection for materials sensitive to light
  • Chemical and thermal resistance
Ideal for microchemical work. Features wide mouth facilitates pipete filling and mixing.
The flasks are supplied with larger interchangeable glass stoppers
Specifications for Product Family
Specifications
  • Catalog No:
  • 66025-682
  • 22877-016
  • 22877-018
  • 77777-081
  • 66025-672
  • 77777-083
  • 33500-516
  • 33500-520
  • 33500-514
  • 33500-518
  • Environmentally Preferable:
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Environmental certification:
  • I-RECs, ISO 14001 - Monterrey
  • I-RECs, ISO 14001 - Monterrey
  • I-RECs, ISO 14001 - Monterrey
  • I-RECs, ISO 14001 - Monterrey
  • I-RECs, ISO 14001 - Monterrey
  • I-RECs, ISO 14001 - Monterrey
  • I-RECs, ISO 14001 - Monterrey
  • I-RECs, ISO 14001 - Monterrey
  • I-RECs, ISO 14001 - Monterrey
  • I-RECs, ISO 14001 - Monterrey
Specifications
Product Family Options
Product Information
  • Capacity
    Stopper No.
    O.D.×H
    Tolerance
    Availability
    Price
  • 66025-682
    Heavy Duty Flasks
    Heavy Duty Flasks
    10 ml
    13
    48 x 135 mm
    ±0.02 ml
    Log in for availability
    Log in for availability
    Case (6UN)
    Case (6UN) - $7,460.00
    $7,460.00
    Case (6UN)
       $7,460.00
    Supplier Number: 5635-10
    Specifications:
    Product available on GSA Advantage®
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    66025-682
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    I-RECs, ISO 14001 - Monterrey
    Product available on GSA Advantage®
      Low Manufacturing Impact
  • 22877-016
    Heavy Duty Flasks
    Heavy Duty Flasks
    25 ml
    13
    40 x 100 mm
    ±0.03 ml
    Log in for availability
    Log in for availability
    Case (6UN)
    Case (6UN) - $7,460.00
    $7,460.00
    Case (6UN)
       $7,460.00
    Supplier Number: 5635-25
    Specifications:
    Product available on GSA Advantage®
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    22877-016
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    I-RECs, ISO 14001 - Monterrey
    Product available on GSA Advantage®
      Low Manufacturing Impact
  • 22877-018
    Heavy Duty Flasks
    Heavy Duty Flasks
    50 ml
    13
    51 x 130 mm
    ±0.05 ml
    Log in for availability
    Log in for availability
    Case (6UN)
    Case (6UN) - $7,770.00
    $7,770.00
    Case (6UN)
       $7,770.00
    Supplier Number: 5635-50
    Specifications:
    Product available on GSA Advantage®
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    22877-018
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    I-RECs, ISO 14001 - Monterrey
    Product available on GSA Advantage®
      Low Manufacturing Impact
  • 77777-081
    Heavy Duty Flasks
    Heavy Duty Flasks
    100 ml
    16
    66 x 160 mm
    ±0.08 ml
    Log in for availability
    Log in for availability
    Case (6UN)
    Case (6UN) - $11,860.00
    $11,860.00
    Case (6UN)
       $11,860.00
    Supplier Number: 5635-100
    Specifications:
    Product available on GSA Advantage®
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    77777-081
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    I-RECs, ISO 14001 - Monterrey
    Product available on GSA Advantage®
      Low Manufacturing Impact
  • 66025-672
    Heavy Duty Flasks
    Heavy Duty Flasks
    200 ml
    19
    72 x 170 mm
    ±0.10 ml
    Log in for availability
    Log in for availability
    Case (6UN)
    Case (6UN) - $14,290.00
    $14,290.00
    Case (6UN)
       $14,290.00
    Supplier Number: 5635-200
    Specifications:
    Product available on GSA Advantage®
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    66025-672
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    I-RECs, ISO 14001 - Monterrey
    Product available on GSA Advantage®
      Low Manufacturing Impact
  • 77777-083
    Heavy Duty Flasks
    Heavy Duty Flasks
    250 ml
    19
    87 x 180 mm
    ±0.12 ml
    Log in for availability
    Log in for availability
    Case (6UN)
    Case (6UN) - $14,970.00
    $14,970.00
    Case (6UN)
       $14,970.00
    Supplier Number: 5635-250
    Specifications:
    Product available on GSA Advantage®
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    77777-083
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    I-RECs, ISO 14001 - Monterrey
    Product available on GSA Advantage®
      Low Manufacturing Impact
  • 33500-516
    Low Actinic, Heavy Duty Flasks
    Low Actinic, Heavy Duty Flasks
    25 ml
    13
    40 x 100 mm
    ±0.03 ml
    Discontinued
    Supplier Number: -
    Specifications:
    Product available on GSA Advantage®
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    33500-516
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    I-RECs, ISO 14001 - Monterrey
    Product available on GSA Advantage®
      Low Manufacturing Impact
  • 33500-520
    Low Actinic, Heavy Duty Flasks
    Low Actinic, Heavy Duty Flasks
    50 ml
    13
    51 x 130 mm
    ±0.05 ml
    Discontinued
    Supplier Number: -
    Specifications:
    Product available on GSA Advantage®
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    33500-520
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    I-RECs, ISO 14001 - Monterrey
    Product available on GSA Advantage®
      Low Manufacturing Impact
  • 33500-514
    Low Actinic, Heavy Duty Flasks
    Low Actinic, Heavy Duty Flasks
    100 ml
    16
    66 x 160 mm
    ±0.08 ml
    Discontinued
    Supplier Number: -
    Specifications:
    Product available on GSA Advantage®
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    33500-514
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    I-RECs, ISO 14001 - Monterrey
    Product available on GSA Advantage®
      Low Manufacturing Impact
  • 33500-518
    Low Actinic, Heavy Duty Flasks
    Low Actinic, Heavy Duty Flasks
    250 ml
    19
    87 x 180 mm
    ±0.12 ml
    Discontinued
    Supplier Number: -
    Specifications:
    Product available on GSA Advantage®
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    33500-518
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    I-RECs, ISO 14001 - Monterrey
    Product available on GSA Advantage®
      Low Manufacturing Impact