VWR® Vortex Mixers
Acerca de este artículo
Fixed high speed mixing and variable speed models.
- Five year warranty for parts and labor
- All models include a cup head and three inch platform
- Manufactured in the USA
VWR® Fixed Speed Vortex Mixers start mixing when the cup head is depressed. Speed is a full 3200 rpm to provide vigorous mixing of samples.
VWR® Analog Vortex Mixers offer variable analog speed control with continuous or touch mode operation. Control allows low rpm start up for gentle shaking or high speed mixing for vigorous vortexing of samples. Two modes of operation; continuous mode when using optional accessory attachments or touch mode which activates mixing when depressing the cup head.
VWR® Digital Vortex Mixers are ideal for applications that demand repeatable results. Microprocessor control maintains set speed for strong, consistent mixing action. Touch pad controls and independent LED displays for speed and time. Timer will display elapsed time or when programmed to a user defined time limit, the unit will shut off when time reaches zero.Two modes of operation; continuous mode when using the optional accessory attachments, or touch mode which activates mixing when depressing the cup head.
VWR® Pulsing Vortex Mixers are ideal for glass bead cell disruption/homogenization and provide a powerful pulsing vortex action.Capable of complete cell disruption of samples in only minutes.Unique pulsing action mixes at maximum speed for 30 seconds and rests for 10 seconds reducing heat generation while providing more effective mixing and disruption. Displayed time counts up during continuous operation and counts down during timed runs. Also includes a stainless steel microtube holder with built-in cup head.
: TUV listed. RoHS compliant.
: All models include a cup head, three-inch head with cover, and a 72" (183 cm), 3-wire cord and plug. Pulsing Vortex Mixer includes a stainless steel microtube holder with built-in cup head and holds up to (12) 1.5 to 2 ml microtubes.
Vortex Mixers with NIST traceable calibration certificate provided by an ISO/IEC 17025 accredited laboratory. Multiple data points within the speed range and time function are reported on the certificate with the associated uncertainties.
5 Options Available Below
Product Details & Documents
Fixed high speed mixing and variable speed models.
- Five year warranty for parts and labor
- All models include a cup head and three inch platform
- Manufactured in the USA
VWR® Fixed Speed Vortex Mixers start mixing when the cup head is depressed. Speed is a full 3200 rpm to provide vigorous mixing of samples.
VWR® Analog Vortex Mixers offer variable analog speed control with continuous or touch mode operation. Control allows low rpm start up for gentle shaking or high speed mixing for vigorous vortexing of samples. Two modes of operation; continuous mode when using optional accessory attachments or touch mode which activates mixing when depressing the cup head.
VWR® Digital Vortex Mixers are ideal for applications that demand repeatable results. Microprocessor control maintains set speed for strong, consistent mixing action. Touch pad controls and independent LED displays for speed and time. Timer will display elapsed time or when programmed to a user defined time limit, the unit will shut off when time reaches zero.Two modes of operation; continuous mode when using the optional accessory attachments, or touch mode which activates mixing when depressing the cup head.
VWR® Pulsing Vortex Mixers are ideal for glass bead cell disruption/homogenization and provide a powerful pulsing vortex action.Capable of complete cell disruption of samples in only minutes.Unique pulsing action mixes at maximum speed for 30 seconds and rests for 10 seconds reducing heat generation while providing more effective mixing and disruption. Displayed time counts up during continuous operation and counts down during timed runs. Also includes a stainless steel microtube holder with built-in cup head.
: TUV listed. RoHS compliant.
: All models include a cup head, three-inch head with cover, and a 72" (183 cm), 3-wire cord and plug. Pulsing Vortex Mixer includes a stainless steel microtube holder with built-in cup head and holds up to (12) 1.5 to 2 ml microtubes.
Vortex Mixers with NIST traceable calibration certificate provided by an ISO/IEC 17025 accredited laboratory. Multiple data points within the speed range and time function are reported on the certificate with the associated uncertainties.
Specifications for Product Family
Especificaciones
- Catalog No:
- 10153-834
- 10153-838
- 10153-842
- 10153-812
- 10153-816
- Environmentally Preferable:
- Waste Reducing
- Waste Reducing
- Waste Reducing
- Waste Reducing
- Waste Reducing
- Shipping weight:
- 10 lbs.
- 10 lbs.
- 10 lbs.
- 10 lbs.
- 10 lbs.
- L×W×H:
- 7×4.4×5.3"
- 7×4.4×5.3"
- 7×4.4×5.3"
- 7×4.4×5.3"
- 7×4.4×5.3"
- Duty rating:
- Intermittent Duty
- Intermittent Duty
- Intermittent Duty
- Intermittent Duty
- Intermittent Duty
- Environmental certification:
- RoHS
- RoHS
- RoHS
- RoHS
- RoHS
Especificaciones
Product Family Options
Vortex Mixers
Información del producto
- ElectricalOrbitSpeedAvailabilityPrice
- 10153-834VWR® Fixed Speed Vortex MixerVWR® Fixed Speed Vortex Mixer120 V, 50/60 Hz, 1.2 A, 135 W4.9 mm3200 rpmLog in Para disponibilidadEach (1UN)$6,290.26Especificaciones:Product available on GSA Advantage®
Waste Reducing More
Specifications
Cat. No.10153-834Environmentally PreferableWaste ReducingShipping weight10 lbs.L×W×H7×4.4×5.3"Duty ratingIntermittent DutyEnvironmental certificationRoHS - 10153-838VWR® Analog Vortex MixerVWR® Analog Vortex Mixer120 V, 50/60 Hz, 1.2 A, 135 W4.9 mm300 - 3200 rpmLog in Para disponibilidadEach (1UN)$7,682.57Especificaciones:Product available on GSA Advantage®
Waste Reducing More
Specifications
Cat. No.10153-838Environmentally PreferableWaste ReducingShipping weight10 lbs.L×W×H7×4.4×5.3"Duty ratingIntermittent DutyEnvironmental certificationRoHS - 10153-842VWR® Digital Vortex MixerVWR® Digital Vortex Mixer120 V, 50/60 Hz, 1.2 A, 135 W4.9 mm500 - 3000 rpmLog in Para disponibilidadEach (1UN)$12,188.51Especificaciones:Product available on GSA Advantage®
Waste Reducing More
Specifications
Cat. No.10153-842Environmentally PreferableWaste ReducingShipping weight10 lbs.L×W×H7×4.4×5.3"Duty ratingIntermittent DutyEnvironmental certificationRoHS - 10153-812VWR® Pulsing Vortex MixerVWR® Pulsing Vortex Mixer120 V, 50/60 Hz, 1.2 A, 135 W2.5 mm500 - 3000 rpmLog in Para disponibilidadEach (1UN)$17,146.81Especificaciones:Product available on GSA Advantage®
Waste Reducing More
Specifications
Cat. No.10153-812Environmentally PreferableWaste ReducingShipping weight10 lbs.L×W×H7×4.4×5.3"Duty ratingIntermittent DutyEnvironmental certificationRoHSProduct available on GSA Advantage®Waste Reducing
Vortex Mixers with NIST Traceable Certificates
Información del producto
- ElectricalOrbitSpeedAvailabilityPrice
- 10153-816VWR® Digital Vortex Mixer with NIST CertificateVWR® Digital Vortex Mixer with NIST Certificate120 V, 50/60 Hz, 1.2 A, 135 W4.9 mm500 - 3000 rpmLog in Para disponibilidadEach (1UN)$15,984.84Especificaciones:Product available on GSA Advantage®
Waste Reducing More
Specifications
Cat. No.10153-816Environmentally PreferableWaste ReducingShipping weight10 lbs.L×W×H7×4.4×5.3"Duty ratingIntermittent DutyEnvironmental certificationRoHSProduct available on GSA Advantage®Waste Reducing
Accessories for tubes and plates
Información del producto
- Catalog #DescriptionAvailabilityPrice
Other accessories
Información del producto
- Catalog #DescriptionAvailabilityPrice
Accessories
Información del producto
- Catalog #AvailabilityPrice
- Rubber head cover, 76 mm (3")
Información del producto
- Catalog #AvailabilityPrice
- Tube holder, 20 to 25 mm
Información del producto
- Catalog #AvailabilityPrice
- Head with rubber cover, 76 mm (3")
Información del producto
- Catalog #AvailabilityPrice
- Flat foam insert, 160 mm (6.3") diameter
Información del producto
- Catalog #AvailabilityPrice
- Microplate holder
Información del producto
- Catalog #AvailabilityPrice
- Single tube holder adapter for Vortex GenieSingle tube holder adapter for Vortex Genie12621-252Log in Para disponibilidadEach (1UN) - $2,153.74
Información del producto
- Catalog #AvailabilityPrice
- Single tube holder
Información del producto
- Catalog #AvailabilityPrice
- Microtube holder
Información del producto
- Catalog #AvailabilityPrice
- Insert retainer
Información del producto
- Catalog #AvailabilityPrice
- Single tube holder accessory for 18×1.5 - 2 ml tubesSingle tube holder accessory for 18×1.5 - 2 ml tubes12620-896Log in Para disponibilidadEach (1UN) - $2,081.85
Información del producto
- Catalog #AvailabilityPrice
- Tube holder kit with insert retainer
Información del producto
- Catalog #AvailabilityPrice
- Head, 76 mm (3")
Información del producto
- Catalog #AvailabilityPrice
- Tube holder, 14 to 19 mm