Orders LinkContactUsLinkComponent
3B Scientific® Kidney Section Models
3B Scientific® Kidney Section Models
Kidney Section, 24, 1, 21 cm, 34 cm, 10 cm
Catalog # 470103-260
Supplier #:  1000296
3B Scientific® Kidney Section Models
Catalog # 470103-260
3B Scientific® Kidney Section Models
3B Scientific® Kidney Section Models
Kidney Section, 24, 1, 21 cm, 34 cm, 10 cm
Price
$6,449.38
Log in for availability
Each (1UN)
Product Details & Documents
These kidney section models are colorful and anatomically accurate.
  • Model Mounted on Baseboard
  • Users' Manual Included
  • Models Depict Right Kidney, Nephrons and Blood Vessels, or Renal Corpuscle
  • Kidney 3 Times Larger than Life-Size
  • Includes 3B Smart Anatomy
They depict a longitudinal section of the right kidney, a detailed view of the nephrons and blood vessels, or a renal corpuscle. The models show the important structures of the human kidney for student and patient education. The kidney is three times larger than life size, the nephron section is approximately 125 times larger than life size, and the corpuscle is 700 times larger than life size.
Each kidney section model is mounted on a baseboard and comes with a users' manual.
Accessories information: For information about 3B Smart Anatomy, please see the document attached on this page.
Specifications
  • Description:
    Kidney Section
  • Height:
    10 cm
  • Length:
    34 cm
  • No. of Keyed Items:
    24
  • Width:
    21 cm
  • No. of Parts:
    1