Orders LinkContactUsLinkComponent
Cryoscope Standards, Advanced Instruments
Cryoscope Standards, Advanced Instruments
Certificates
Cryoscope Standards, Advanced Instruments
2 Options Available Below
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Cryoscope standards are used to calibrate and/or check the performance of a cryoscope.
Standards should only be used in accordance with the cryoscope being calibrated. Refer to the specific instrument's user guide for calibration instructions. These standards are prepared from salts. 2-Hydroxybiphenyl Sodium Salt Tetrahydrate, 0.01%, is added as a preservative.
Product Family Options
Información del producto
  • Availability
    Price
  • 52198-144
    Cryoscope Standard 422 m°H, 110 mL
    Cryoscope Standard 422 m°H, 110 mL
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (110ml)
    Each (110ml) - $1,945.51
    $1,945.51
    Each (110ml)
       $1,945.51
    Supplier Number: 3LA022
  • 55005-001
    Cryoscope Standard 422 m°H, 10 x 10 mL ampules
    Cryoscope Standard 422 m°H, 10 x 10 mL ampules
    Log in Para disponibilidad
    Log in Para disponibilidad
    Paquete de 10 (10ml)
    Paquete de 10 (10ml) - $4,432.43
    $4,432.43
    Paquete de 10 (10ml)
       $4,432.43
    Supplier Number: 3LA023
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment.