ContactUsLinkComponent
VWR® Advanced Mini Dry Block Heater and Mini Dry Block Heater with Heated Lid, 120 V
VWR® Advanced Mini Dry Block Heater and Mini Dry Block Heater with Heated Lid, 120 V
Supplier:  VWR International
   Sustainable Options Available
Certificates
undefined
VWR® Advanced Mini Dry Block Heater and Mini Dry Block Heater with Heated Lid, 120 V
Supplier:  VWR International
   Sustainable Options Available
Certificates
Specifications
  • Catalog No:
  • 10153-352
  • 10153-348
  • 10153-318
  • 10153-356
  • Environmentally Preferable:
  • Waste Reducing
  • Waste Reducing
  • Waste Reducing
  • Waste Reducing
  • Intervalo de temperaturas:
  • Ambient +5 to 100°C
  • Ambient +5 to 100°C
  • Ambient +5 to 100°C
  • Ambient +5 to 100°C
  • Uniformidad de la temperatura:
  • +/- 0.2°C
  • +/- 0.2°C
  • +/- 0.2°C
  • +/- 0.2°C
  • Pr×An×Al:
  • 160×127×127 mm (6.3×5×4.85")
  • 160×127×127 mm (6.3×5×4.85")
  • 160×127×127 mm (6.3×5×4.85")
  • 160×127×127 mm (6.3×5×4.85")
  • Peso al enviarse:
  • 1.6 kg (3.5 lbs.)
  • 1.6 kg (3.5 lbs.)
  • 1.6 kg (3.5 lbs.)
  • 1.6 kg (3.5 lbs.)
  • Certificación ambiental:
  • RoHS Compliant
  • RoHS Compliant
  • RoHS Compliant
  • RoHS Compliant
Specifications
VWR® Mini Modular Blocks
Product Information
Frequently Bought Together