Inoculation Loops, Calibrated, Disposable
Flexible (Soft) Plastic Loop, 10 μl, Light Blue, 200/box
Supplier: COPAN DIAGNOSTICS INC
: COP-S10
Jump to:
Product Details & Documents
Ultra smooth loop surfaces allows easy streaking
No rough plastic edges, flashing or burrs on the loop head means smooth, problem-free planting and streaking of cultures. Loops do not cut or gouge the agar surface during streaking. Free of lubricants, oils and electrostatic charges enabling consistent wetting and complete liquid transfer. Hexagonal loop shaft improves grip, assists orientation and makes diluting and streaking easy
: Loop accuracy certified using Evans Blue Dye Method. Certification of Calibration in every box.
: Packaged as 25 loops per bag and 1,000 loops per box. Zip-lock closure reseals the pack between use to avoid contamination.
Certificates
Buscar por número de loteEspecificaciones
- Description:Flexible (Soft) Plastic Loop
- Size:10 μl
- Packaging:200/box
- Handle Color:Light Blue