Antibiotic Moisture Content Flasks, Kimble Chase, DWK Life Sciences
Catalog # KT881700-0000
Supplier: DWK LIFE SCIENCES S. DE R.L. DE MX
Specifications
- Capacidad:2.2 ml
- Referencia:KT881700-0000
- ST Joint Size:10/10
- Nº de proveedor:881700-0000
Specifications
About this item
For determining moisture loss on drying in tablets, troches, or capsules.
Manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements.