Serialized and Certified Automatic Zero Burets with PTFE Stopcock and Reservoir Bottle, Class A, Kimble Chase
Supplier: DWK LIFE SCIENCES S. DE R.L. DE MX
Certificates
About this item
KIMAX® precision bore automatic burets are used in applications requiring the highest degree of precision and accuracy for volumetric analysis
Manufactured to the specifications found in ASTM E694. Calibrated to the accuracy requirements found in ASTM Specification E542. Manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements.
These are ideal for repeat titrations requiring traceable volumetric accuracy or when the titrant should not be handled.Packed complete with a reservoir bottle, U-shaped drying tube, vented connecting tube, rubber squeeze bulb, #1 single-holed rubber stopper, PTFE stopcock plug and 1/4 inch ID rubber tubing.
Certifications: Supplied with a Certificate of Graduation Accuracy.
Related Information
Frequently Bought Together