Orders LinkContactUsLinkComponent
VWR® Ice Pans
VWR® Ice Pans
VWR® Ice Pans
Supplier:  VWR International
Certificates
VWR® Ice Pans
5 Options Available Below
Product Details & Documents
VWR® Ice Pans are manufactured from polyurethane foam to ensure good insulating properties.
  • Polyurethane foam construction
  • pouring lip for easy draining
  • Lids can be stacked beneath pans
Ice Pans are suitable for use with Dry-Ice with Isopropyl Alcohol, Water-Ice with salt slurry, or liquid nitrogen. The operating temperature range is -76 to 70°C (-104 to 158°F). All pans feature a pouring lip for easy draining. VWR® Ice Pans are available with or without lids. The lids, which are unbreakable in normal use, help to reduce evaporation levels. Lids can also be stacked below the pans for convenient, space-saving storage.
Documents
Product Family Options
VWR® Ice Pans without Lid
Información del producto
  • Capacity
    Color
    Width
    Depth
    Height
    Availability
    Price
  • Small
    1 L (0.26 gal.)
    Red
    215 mm (8.5")
    188 mm (7.4")
    118 mm (4.6")
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (1UN)
    Each (1UN) - $796.31
    $796.31
    Each (1UN)
       $796.31
  • Medium
    4 L (1.05 gal.)
    Red
    380 mm (14.9")
    260 mm (10.2")
    155 mm (6.1")
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (1UN)
    Each (1UN) - $1,242.45
    $1,242.45
    Each (1UN)
       $1,242.45
VWR® Ice Pans with Lid
Información del producto
  • Capacity
    Color
    Width
    Depth
    Height
    Availability
    Price
  • Small
    1 L (0.26 gal.)
    Blue
    215 mm (8.5")
    188 mm (7.4")
    118 mm (4.6")
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (1UN)
    Each (1UN) - $977.56
    $977.56
    Each (1UN)
       $977.56
  • Medium
    4 L (1.05 gal.)
    Black
    380 mm (14.9")
    260 mm (10.2")
    155 mm (6.1")
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (1UN)
    Each (1UN) - $1,667.26
    $1,667.26
    Each (1UN)
       $1,667.26
  • Medium
    4 L (1.05 gal.)
    Blue
    380 mm (14.9")
    260 mm (10.2")
    155 mm (6.1")
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (1UN)
    Each (1UN) - $1,667.26
    $1,667.26
    Each (1UN)
       $1,667.26