Science Take-Out® Brains Under The Influence
Proveedor: SCIENCE TAKE-OUT, LLC
Certificados
Acerca de este artículo
What Factors Affect Blood Alcohol Concentration?
- Available As A Single Kit With Materials For 1 Student Or Group
- Unassembled 10 Pack For Additional Savings
- No Additional Equipment Needed
Conduct simulated blood alcohol tests and learn about alcohol poisoning and the effects of binge drinking on teens’ brains. Collect and graph data on the effect of alcohol consumption on blood alcohol concentration. Match the information from six mini-cases with the parts of the brain affected by alcohol. Learn about research that links teen binge drinking to irreversible brain damage. This complete experiment setup contains all materials and does not require lab equipment.
Frequently Bought Together