Orders LinkContactUsLinkComponent
Science Take-Out® Cell Communication
Science Take-Out® Cell Communication
Science Take-Out® Cell Communication
Certificates
Science Take-Out® Cell Communication
Science Take-Out® Cell Communication
2 Options Available Below
Product Details & Documents
Create A Model To Illustrate Cell Communication.
  • Available As A Single Kit With Materials For 1 Student Or Group
  • Unassembled 10 Pack For Additional Savings
  • No Additional Equipment Needed
Create a model to explore how cells send, receive, and respond to signals. Use this model to illustrate cell communication involved in regulating blood sugar level and in nerve cell action. This complete "dry lab" activity contains all required materials.
Product Family Options
Información del producto
  • No. of Lab Groups
    Availability
    Price
  • 470118-348
    Cell communication single kit
    Cell communication single kit
    1
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (1UN)
    Each (1UN) - $271.00
    $271.00
    Each (1UN)
       $271.00
    Supplier Number: STO-133
  • 470118-382
    Cell communication unassembled 10 pack
    Cell communication unassembled 10 pack
    10
    Log in Para disponibilidad
    Log in Para disponibilidad
    Paquete de 10
    Paquete de 10 - $2,120.00
    $2,120.00
    Paquete de 10
       $2,120.00
    Supplier Number: STO-133U