Periscope
Periscope
Catalog # 470117-800
Supplier: UNITED SCIENTIFIC SUPPLIES
Número de proveedor #: PSCOPE
Jump to:
Product Details & Documents
Demonstrate how periscopes in submarines work.
- Sturdy design
- Mirrors securely held on each end
Inspired by naval periscopes used on submarines, our handheld periscope is designed to show students the reflective qualities of light. Our version consists of an outer case, with mirrors at each end set parallel to each other at a 45-degree angle.
Especificaciones
- Description:Periscope