ContactUsLinkComponent
The SpillNot™ Centripetal Force Demonstrator
The SpillNot™ Centripetal Force Demonstrator
Catalog # 470117-798
undefined
The SpillNot™ Centripetal Force Demonstrator
Catalog # 470117-798
Supplier Number:  SPNT01
Specifications
  • Descripción:
    The SpillNot™
  • Referencia:
    470117-798
  • Nº de proveedor:
    SPNT01
Specifications
About this item
Vividly demonstrate centripetal forces with this exciting apparatus.
  • Non-slip base
  • Easy to grab handle for spinning
  • SpillNot™ holds one glass securely
  • SpillNot Plus™ holds up to three glasses securely
The design of the SpillNot™ uses the principles of Newtonian physics to keep the liquid in the container. The flexible handle and the location of the suspension point above the center of the tray ensure that the forces on the liquid are always directed towards the supporting tray, even when the tray is in motion. The sideways forces that are usually responsible for spills are not transmitted to the tray by the flexible handle and the radial forces generated during swinging motion only pin the liquid into the container more firmly. The larger design of the SpillNot™ Plus holds three glasses at once.
An inexpensive demonstration to intrigue students and challenge their analytical powers!
Delivery: Includes activity guide.