Polarizing Film, 6 × 6”
Proveedor: UNITED SCIENTIFIC SUPPLIES
Certificados
Acerca de este artículo
Demonstrate how polarizing filters affect light.
- Comes with a support stand
- Comes as pairs to filter out light
Unmounted 6” x 6” film is an effective yet economical tool for demonstrating polarization of light. When the two filters are placed 90 degrees to each other, they block out a majority of the incoming light.
Frequently Bought Together