VWR® Basic Frosted Microscope Slide without Tissue Paper
Slides, 1.0 mm, 75×25 mm (3×1")
Supplier: VWR International
VWR® Basic Frosted Microscope Slide without Tissue Paper
Catalog # 89085-399
Supplier: VWR International
Jump to:
Product Details & Documents
VWR® Frosted Microscope Slides without Tissue Paper are specially designed with consistent and clean surfaces for staining experiments.
- Consistent, clean surfaces
- Frosted edge for marking
They are ideal for histology, hematology, and microbiology applications. Slides feature frosted areas on both sides of one end for convenient marking. A fine polished edge and 45° clipped corners promote ease and safety in handling. Slides are packaged 72 per pack, 20 packs per case.
Documents
Especificaciones
- Description:Slides
- L×W:75×25 mm (3×1")
- Thickness:1.0 mm