Orders LinkContactUsLinkComponent
Cardboard Cryogenic Vial Boxes
Supplier:   HEATHROW SCIENTIFIC
Cardboard Cryogenic Vial Boxes
Certificates
Cardboard Cryogenic Vial Boxes
Cardboard Cryogenic Vial Boxes
2 Options Available Below
Product Details & Documents
Boxes for storage of small samples and tubes.
  • Made from cardboard
  • Colors allow for easy identification and sorting purpose
  • Store in LN₂, or in mechanical freezers
  • Bottom vent holes to allow quicker circulation and removal of cold air, vapor or LN₂
  • Fits 1.5/2.0 ml tubes
Each assorted pack includes four each of blue, red, and yellow. Bottom footprint is 133×133 mm.
Product Family Options
Información del producto
  • Color
    Dimensions
    Availability
    Price
  • 490007-640
    Vial box and lid
    Vial box and lid
    Assorted (red, blue, and yellow)
    133×133×50 mm
    Log in Para disponibilidad
    Log in Para disponibilidad
    Log in Para disponibilidad
    Paquete de 12
    Paquete de 12 - $1,375.10
    $1,375.10
    Log in Para disponibilidad
    Case de 4 (12UN)
    Case de 4 (12UN) - $1,498.86
    $1,498.86
    Paquete de 12
       $1,375.10
    Case de 4 (12UN)
       $1,498.86
    Supplier Number: HS2860C
  • 490007-642
    100 cell partition (10×10 format)
    100 cell partition (10×10 format)
    White
    129×129×381 mm
    Log in Para disponibilidad
    Log in Para disponibilidad
    Log in Para disponibilidad
    Paquete de 12
    Paquete de 12 - $1,102.68
    $1,102.68
    Log in Para disponibilidad
    Case de 40 (12UN)
    Case de 40 (12UN) - $1,201.92
    $1,201.92
    Paquete de 12
       $1,102.68
    Case de 40 (12UN)
       $1,201.92
    Supplier Number: HS2860E