Orders LinkContactUsLinkComponent
EZ-CFU™ One Step, Microbiologics
Kocuria rhizophila, ATCC® 9341™*
Catalog # 10142-632
 :  0688Z
EZ-CFU™ One Step, Microbiologics
Catalog # 10142-632
CAS Number:  
EZ-CFU™ One Step, Microbiologics
EZ-CFU™ One Step, Microbiologics
Kocuria rhizophila, ATCC® 9341™*
CAS Number:  
Price
$927.49
Log in Para disponibilidad
Each (1UN)
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
  • El artículo requiere control de temperatura para almacenamiento y entrega con cargos adicionales. No es elegible para devolución debido a preocupaciones de seguridad y calidad. Considere los requisitos antes de comprar.
  • Return Policy
Product Details & Documents
The Growth Promotion Test (GPT) is an important function in the United States (USP), European (Ph. Eur.) and Japanese (JP) Pharmacopeias. EZ-CFU™ One Step quantitative QC microorganism preparations are used to perform Growth Promotion Tests of culture media with ease. No dilutions are required. EZ-CFU™ One Step is designed to deliver 10-100 CFU with each 0.1 mL inoculum. Each 2 mL vial of hydrated suspension offers 19 inocula.
EZ-CFU™ One Step kits include 20 lyophilized pellets of a single quantitative QC microorganism, 10 vials of hydrating fluid (2 mL per vial) and Peel-Off Certificate of Assay
These tests have an eight hour stability time, with extended time and flexibility to perform tests. The Clostridium perfringens and the Bacteroides fragilis tests must be used within 30 minutes of hydration.
Tests require less prep time because no dilutions are required.
 : To receive this product, customers must have a valid ATCC End User Licensing Agreement in place with Microbiologics. Registration for this agreement is only required once, per location during the first order. Instructions for how to register can be downloaded from the product page. Go to the "Related Information" box, click on "Additional Product Details", then "Instructions for End User Registration with Microbiologics".
The ATCC Licensed Derivative Emblem, the ATCC Licensed Derivative word mark, and the ATCC catalog marks are trademarks of ATCC. Microbiologics Inc. is licensed to use these trademarks and to sell products derived from ATCC® cultures.
Especificaciones
Especificaciones
  • Description:
    Kocuria rhizophila
  • ATCC No.:
    ATCC® 9341™*
Especificaciones
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment.