Orders LinkContactUsLinkComponent
Ward's® Blood Typing Model
Supplier:   Avantor
Ward's® Blood Typing Model
Supplier:  Avantor
Certificates
Ward's® Blood Typing Model
Ward's® Blood Typing Model
2 Options Available Below
Product Details & Documents
Ward's Blood Typing Model gives students a visual interpretation of a difficult concept.
  • Kit Explores Antigen-Antibody Interactions
  • Lab Functions as Demonstration or Class Activity
  • Estimated Lab Time: 20 to 30 Minutes
  • Clear Demonstration of Complex Concept
It recreates the antigen-antibody interactions occurring on the surface of red blood cells.
Using the model, teachers can demonstrate the antigen-antibody reactions that occur at the molecular level including blood-typing reactions, successful and unsuccessful blood transfusion reactions, and Rh incompatibility.
 : This model includes two erythrocytes and one pair each of A, B, and Rh antigens and antibodies. The Class Size Kit contains six sets of material contained in the demonstration set.
Product Family Options
Product Information
  • No. of Lab Groups
    Availability
    Price
  • 470001-896
    Blood Typing Demonstration Model, Demo Size
    Blood Typing Demonstration Model, Demo Size
    1
    Log in for availability
    Log in for availability
    Each (1UN)
    Each (1UN) - $954.60
    $954.60
    Each (1UN)
       $954.60
  • 470001-898
    Blood Typing Demonstration Model, Class Size
    Blood Typing Demonstration Model, Class Size
    6
    Log in for availability
    Log in for availability
    Each (1UN)
    Each (1UN) - $5,185.80
    $5,185.80
    Each (1UN)
       $5,185.80