ContactUsLinkComponent
Avanti™ JXN Series High Performance Centrifuges, Beckman Coulter®
Avanti™ JXN Series High Performance Centrifuges, Beckman Coulter®
Catálogo # 10119-926
Proveedor:  BECKMAN COULTER
Product available on GSA Advantage®
undefined
Avanti™ JXN Series High Performance Centrifuges, Beckman Coulter®
Catálogo # 10119-926
Proveedor:  BECKMAN COULTER
Número de proveedor:  B38619
Product available on GSA Advantage®
Especificaciones
  • Descripción:
    Avanti™ JXN-26 High Performance Centrifuge
  • Referencia:
    10119-926
  • Velocidad máxima:
    26,000 rpm
  • RCF máximo:
    81770×g
  • Capacidad máxima:
    6 L
  • Nº de proveedor:
    B38619
Especificaciones
Acerca de este artículo
The Avanti™ JXN Series of High Performance Centrifuges offer the functionality, design level, performance, and versatility to meet any laboratory's needs.
  • Foot pedal for hands-free operation
  • Remote monitoring and control through Molecule App
  • Friction Reduction System (FRS)
  • Dynamic Rotor Inertia Check (DRIC)
  • BioSafe options
The Avanti™ JXN Series features a low work-surface height, a door that swings up and out of the way for easy rotor chamber access, and large digital readouts
Additional standard features include a high-torque drive that shortens cycle times, BioSafe options, and more. With password protection, e-signature which permits users to electronically add a signature and a note (if desired), numerous stored protocols, and networking across multiple instruments, data management options are ideal for GMP environment or shared laboratories. The Avanti™ JXN Series offers ergonomic design, intuitive interface, remote control and monitoring, safety and sustainability, easy-to-use data management, and versatility across applications.
The intuitive interface, with a 15" LCD screen, provides a view of set and actual run conditions simultaneously and rotor selection by name. The MobileFuge app allows users to access data and user management and remote monitoring and control via networks and mobile devices. Remote control and monitoring enables users to set and monitor run parameters remotely with mobile devices and receive alerts and diagnostics remotely.
Información de pedido: Rotors and optional accessories sold separately. For high performance centrifuge bottles and tubes, contact your Avantor sales representative.