Blood Typing Trays, MED-TEK
Catalog # 470190-886
Supplier: MED-TEK CORPORATION
Specifications
- Descripción:Blood Typing Trays
- Referencia:470190-886
- Nº de proveedor:701
Specifications
About this item
These styrene trays contain depression wells to perform ABO and Rh blood-typing.
- Well style trays contain liquids
- Clear plastic shows agglutination easier
Trays are washable, reusable or disposable.