Ward's® Bulk Simulated Blood
: Avantor
Big enough for all your lab sections.
- Large Quantities
- Each Blood Type Provided in 1 L Bottle
- Convenient Bulk Refills
At the request of our college customers, we now offer these convenient bulk refills for our exclusive Simulated ABO and Rh Blood-Typing Lab Activities.
Instructions for performing blood typing using this product can be found in the teacher's guide and student copy master that accompanied the purchase of the lab activity.
: Each blood type is provided in a 1L bottle. This product requires Ward's Simulated Antisera, available separately.
Frequently Bought Together