Orders LinkContactUsLinkComponent
Science Take-Out® Mark-Recapture Population Sampling
Science Take-Out® Mark-Recapture Population Sampling
Science Take-Out® Mark-Recapture Population Sampling
Certificates
Science Take-Out® Mark-Recapture Population Sampling
Science Take-Out® Mark-Recapture Population Sampling
2 Options Available Below
Product Details & Documents
How Many Spiders Are Hiding In The Leaf Litter?
  • Available As A Single Kit With Materials For 1 Student Or Group
  • Unassembled 10 Pack For Additional Savings
  • No Additional Equipment Needed
How many plastic spiders are hiding in simulated “leaf litter”? Use the mark-recapture population sampling technique to estimate the number of organisms in a population. Capture, mark and release a random sample of spider in your "leaf litter". Recapture a second random sample of spiders and use a mathematical equation to estimate the total number of animals in the population. This complete "dry lab" activity contains all required materials.
Documents
Product Family Options
Información del producto
  • No. of Lab Groups
    Availability
    Price
  • 470017-740
    Mark-Recapture population sampling single kit
    Mark-Recapture population sampling single kit
    1
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (1UN)
    Each (1UN) - $217.00
    $217.00
    Each (1UN)
       $217.00
    Supplier Number: 111
  • 470017-780
    Mark-Recapture population sampling unassembled 10 pack
    Mark-Recapture population sampling unassembled 10 pack
    10
    Log in Para disponibilidad
    Log in Para disponibilidad
    Paquete de 1
    Paquete de 1 - $1,420.00
    $1,420.00
    Paquete de 1
       $1,420.00
    Supplier Number: 111U