Science Take-Out® From DNA To Protein; Structure And Function
Proveedor: SCIENCE TAKE-OUT, LLC
Certificados
Acerca de este artículo
Create A Protein Model.
- Available As A Single Kit With Materials For 1 Student Or Group
- Unassembled 10 Pack For Additional Savings
- No Additional Equipment Needed
What do genes do? Model how the coded information in genes results in proteins that perform specific functions in the body. Transcribe the DNA code to make a messenger RNA code. Translate the RNA code to make a sequence of amino acids in a protein. Create a bead and chenille stem model of a folded protein. Explore how protein shapes affect proteins function in your body. This complete "dry lab" activity contains all required materials.
Frequently Bought Together