Orders LinkContactUsLinkComponent
Science Take-Out® Beebops Genetics And Evolution
Science Take-Out® Beebops Genetics And Evolution
Science Take-Out® Beebops Genetics And Evolution
Certificates
Science Take-Out® Beebops Genetics And Evolution
Science Take-Out® Beebops Genetics And Evolution
3 Options Available Below
Product Details & Documents
Create A Baby Beebop!
  • Available As A Single Kit With Materials For 1 Student Or Group
  • Unassembled 10 Pack For Additional Savings
  • No Additional Equipment Needed
Why are Beebop babies in the same family different? Model how sexual reproduction results in offspring with different characteristics. Which Beebop babies are most likely to survive and reproduce? Explore how natural selection affects the frequency of characteristics and genes in the Beebop population. This complete "dry lab" activity contains all required materials.
Documents
Product Family Options
Información del producto
  • No. of Lab Groups
    Availability
    Price
  • 470017-732
    Beebops genetics and evolution single kit
    Beebops genetics and evolution single kit
    1
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (1UN)
    Each (1UN) - $307.00
    $307.00
    Each (1UN)
       $307.00
    Supplier Number: 105
  • 470017-772
    Beebops genetics and evolution unassembled 10 pack
    Beebops genetics and evolution unassembled 10 pack
    10
    Log in Para disponibilidad
    Log in Para disponibilidad
    Paquete de 1
    Paquete de 1 - $2,160.00
    $2,160.00
    Paquete de 1
       $2,160.00
    Supplier Number: 105U
  • 470017-840
    Beebops genetics and evolution unassembled 10 pack refill
    Beebops genetics and evolution unassembled 10 pack refill
    10
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (1UN)
    Each (1UN) - $1,620.00
    $1,620.00
    Each (1UN)
       $1,620.00
    Supplier Number: 105R