Science Take-Out® Yeast Populations
Supplier: SCIENCE TAKE-OUT, LLC
Certificates
About this item
What Environmental Conditions Can Affect Organism Growth?
- Available As A Single Kit With Materials For 1 Student Or Group
- Unassembled 10 Pack For Additional Savings
- No Additional Equipment Needed
Test samples from a simulated yeast population to determine yeast population size, the pH of the environment, and the amount of sugar present. Graph your results and compare them with results from another researcher. This complete experiment setup contains all materials and does not require lab equipment.
Frequently Bought Together