Science Take-Out® Modeling Photosynthesis And Cellular Respiration
Proveedor: SCIENCE TAKE-OUT, LLC
Certificados
Acerca de este artículo
Use Snap Beads To Model The Reactants And Products Of Photosynthesis.
- Available As A Single Kit With Materials For 1 Student Or Group
- Unassembled 10 Pack For Additional Savings
- No Additional Equipment Needed
Create simple snap bead models to illustrate the reactants and products of photosynthesis and cellular respiration. Use review cards to help distinguish between the processes of photosynthesis and cellular respiration. This complete "dry lab" activity contains all required materials.
Frequently Bought Together