GC Autosampler Syringes for Bruker/Varian/Chrompack, Hamilton
Catalog # 10118-410
Supplier: HAMILTON COMPANY
UL Listed
Specifications
- Volumen:10 µl
- Largo:51 mm (2")
- Point style:5
- Referencia:10118-410
- Calibrador:26s
- Modelo:Microliter 701
- Nº de proveedor:202880
- Termination:RN
Specifications
About this item
Hamilton maintains a large catalog of replacement syringes for the most popular GC autosamplers
Hamilton offers one quality syringe option for the Varian 8100 and 8200, four quality syringe options for the Varian Chrompack CP-8400, CP-8410, CP-9019, and CP-9050, and one quality syringe option for the Varian Chrompack CP-9020, and CP-9025 GC Autosamplers.
Our syringes are handmade with an unmatched attention to detail. The manufacturing process and quality assurance procedures ensure that every syringe we sell will provide superior accuracy and precision as well as cycle life. Choose the sub category that is associated with your autosampler manufacturer to find the syringes available for your instrument.
Certifications: UL Listed. CE compliant.
Ordering information: Accessories not included.