ContactUsLinkComponent
Intelli-Sense™ Multi-Speed PVC Blowers, Labconco
Intelli-Sense™ Multi-Speed PVC Blowers, Labconco
  10128-050
Product available on GSA Advantage®
   Energy Efficient
undefined
Intelli-Sense™ Multi-Speed PVC Blowers, Labconco
  10128-050
 :  7183412
Product available on GSA Advantage®
   Energy Efficient
  • Descripción:
    Intelli-Sense™ Multi-Speed PVC Blower
  • Environmentally Preferable:
    Energy Efficient
  • Referencia:
    10128-050
  • Detalles eléctricos:
    115 V, 50/60 Hz
  • Inlet Size:
    12"
  • Outlet Size:
    12"
  • Nº de proveedor:
    7183412
  • Power Capacity:
    1 hp
  • Flow:
    200 - 1325 cfm
Intelli-Sense™ Multi-Speed PVC Blowers handle extremely corrosive atmospheres produced by high order oxidizers such as perchloric acid
They feature a direct drive ECM motor, which consumes one third less energy and is easier and less expensive to maintain than belt drive motors, and a phenolic-coated impeller and housing. Unit comes with control box and 3 speed blower switch. Adjustable over broad range of airflows.
The Intelli-Sense Blower provides an economical alternative to traditional on/off blowers. Users may close the sash and turn the Intelli-Sense Blower switch to standby mode so that the hood operates at a lower velocity yet fulfills minimum air exchange requirements. Lower velocity modes have the added benefit of quieter operation than full velocity mode.
They have a drain connection on the lower portion of the housing so that a washdown system can be utilized. Housings are vacuum formed from unplasticized Type I polyvinyl chloride for complete corrosion protection. Impellers made of injection molded PVDF resist highly corrosive atmospheres, operate quietly and provide optimum delivery. Powder-coated base and weather cover provide long life in extreme weather.
 : Models without Control Box are only for use with Labconco Protector I-S series Fume Hoods.