Legato 100 Single Syringe Infuse Only Syringe Pump, KD Scientific
Catalog # 10117-894
Supplier: HARVARD APPARATUS
cUL Listed
CSA Certified
UL Listed
Specifications
- Precisión:±0.5%
- Velocidad mín. de flujo:1.26 pl/min
- Descripción:Legato 100 Single Syringe Infuse Only Syringe Pump
- Detalles eléctricos:100-220V, 50-60 Hz
- Velocidad de flujo máx.:88.28 ml/min
- Referencia:10117-894
- Nº de proveedor:788100
Specifications
About this item
Infuse only single syringe pump
- Touch pad lock feature, LED light on front panel, USB port, I/O and TTL interface
- Full metal chassis
- Built in syringe table, RS485 interface
- Advanced microstepping techniques
- Continuous mode of operation
- Spill dam
- Programmable
Accomodates syringe sizes 0.5ul to 60 ml. User definable flow rates with selectable target volume or time values. High resolutuion touch screen display. Built in syringe table. Up to 30 lbs of linear force. USB control.
Certifications: UL, CSA, CUL, CE (Global), RoHS, WEEE.
Caution: KD Scientific syringe pumps are for laboratory use only. Not for clinical use.