Corning® Polypropylene Erlenmeyer Flasks, Corning
Catalog # 30617-088
Supplier: CORNING CA
Product available on GSA Advantage®
Specifications
- Intervalo de graduación:20 ml
- Capacidad:125 ml
- Referencia:30617-088
- Grosor:GL-40
- Nº de proveedor:4985P-100
Specifications
About this item
PP, translucent.
- Very good chemical resistance
- Very good temperature resistance
With blue, easy to read graduations. Autoclavable. The flasks feature a stopper bed and can also be sealed with standard ground joint polypropylene stoppers.