Terminator Jaw Crusher, T.M. Engineering
Terminator Jaw Crusher, 591 kg (1300 lbs.)
Catalog # CA10127-248
Supplier: TM ENGINEERING CA
: JCT#1AL ALLOY-1/ALLOY-2
Jump to:
Product Details & Documents
The new Terminator jaw crusher features an opening adjustment on one side, using only two screws
The Terminator allows for the finest media obtainable with a Jaw Crusher, with quicker and simpler adjustment of jaws. Jaws are extra heavy-duty and require less maintenance with automatic lubrication feature, standard. Available Safety (Feeder) Hopper prevents fly-out of rocks while feeding the machine.
New crushing components will outlast previous models.
: Maintenance kits and tools for on-site repairs are available. For more information, contact your Avantor sales representative.
Especificaciones
- Description:Terminator Jaw Crusher
- Weight:591 kg (1300 lbs.)