Head and Face Shield Combinations, 3M™
Head and face protection, with ratchet headgear H8A and 3M™ Clear propionate faceshield W96, Clear
Catalog # 89006-097
Supplier: 3M OCCUPATIONAL HEALTH
Número de proveedor #: 7000127244
Jump to:
Product Details & Documents
A combination head, face, and hearing protection solution
- Combination head, face, and hearing protection solution
- Includes six-point ratchet suspension hard hat
- UVicator™ sensor, metal mesh visor, earmuff, rainshield, and visor seal
- Requires protective eyewear (sold separately)
Combinations include a six-point ratchet suspension hard hat with UVicator™ sensor, metal mesh visor, earmuff, rainshield, and visor seal. Requires protective eyewear, which is sold separately.
Certifications: Items meet ANSI Z87.1-2003 standards.
Especificaciones
- Color:Clear
- Description:Head and face protection, with ratchet headgear H8A and 3M™ Clear propionate faceshield W96