STAR™ DISH Petri Dishes, Semi-Nesting Alpine, Phoenix
25389-032
- Descripción:Semi-Nesting Alpine, Semi-Stacking Petri Dish without Vents, with ISO Mark
- Dimensiones:90 x 15 mm
- Referencia:25389-032
- Nº de proveedor:371
These petri dishes are most suitable for use with high-speed media pouring machines
The recessed lid has a smooth beveled angle allowing the dish to slide easily during automated pouring.