Orders LinkContactUsLinkComponent
Jar, Straight Sided, Polypropylene, Dynalon
Jar, Straight Sided, Polypropylene, Dynalon
Supplier:   DYNALON
Jar, Straight Sided, Polypropylene, Dynalon
Supplier:  DYNALON
Certificates
Jar, Straight Sided, Polypropylene, Dynalon
Jar, Straight Sided, Polypropylene, Dynalon
2 Options Available Below
Product Details & Documents
Straight-sided polypropylene containers are supplied with white polypropylene screw closures
These autoclavable, translucent jars have many laboratory and industrial applications. Lightweight and easy to fill with a maximum operating temperature of 135°C (275°F).
Product Family Options
Información del producto
  • Capacity
    Height
    Ø
    Weight
    Availability
    Price
  • 120 ml (4 oz.)
    60 mm
    70 mm
    0.06875 lbs.
    Log in Para disponibilidad
    Log in Para disponibilidad
    Case (36UN)
    Case (36UN) - $3,378.54
    $3,378.54
    Case (36UN)
       $3,378.54
    Supplier Number: 421135
  • 250 ml (8 oz.)
    66 mm
    89 mm
    0.104 lbs.
    Log in Para disponibilidad
    Log in Para disponibilidad
    Case (36UN)
    Case (36UN) - $4,343.00
    $4,343.00
    Case (36UN)
       $4,343.00
    Supplier Number: 421145