Orders LinkContactUsLinkComponent
Snap Cap Containers, Polystyrene, Dynalon
Snap Cap Containers, Polystyrene, Dynalon
Supplier:   DYNALON
Snap Cap Containers, Polystyrene, Dynalon
Supplier:  DYNALON
Certificates
Snap Cap Containers, Polystyrene, Dynalon
Snap Cap Containers, Polystyrene, Dynalon
3 Options Available Below
Product Details & Documents
Snap Cap Containers are molded of rigid-wall polystyrene with tight fitting polyethylene Snap-On caps
Approximately 16 drams per ounce. Maximum operating temperature for polystyrene is 70 °C.
Excellent for storing dry materials, powders, tablets, etc. The 7 dram size may be used with automatic cell counters for collection of sample and, where venous samples are used, for dilution purposes.
Product Family Options
Product Information
  • Capacity
    Height
    Weight
    Ø
    Availability
    Price
  • 8 ml (3 dram)
    48 mm (without Cap)
    0.00937 lbs.
    17 mm
    Log in for availability
    Log in for availability
    Case (144UN)
    Case (144UN) - $2,575.09
    $2,575.09
    Case (144UN)
       $2,575.09
    Supplier Number: 426364-03
  • 17 ml (5 dram)
    50 mm (without Cap)
    0.01187 lbs.
    22 mm
    Log in for availability
    Log in for availability
    Case (144UN)
    Case (144UN) - $2,631.86
    $2,631.86
    Case (144UN)
       $2,631.86
    Supplier Number: 426364-05
  • 24 ml (7 dram)
    50 mm (without Cap)
    0.01687 lbs.
    26 mm
    Log in for availability
    Log in for availability
    Case (144UN)
    Case (144UN) - $2,484.48
    $2,484.48
    Case (144UN)
       $2,484.48
    Supplier Number: 426364-07