Orders LinkContactUsLinkComponent
VWR® Microdispenser Replacement Kits
Supplier:   VWR International
VWR® Microdispenser Replacement Kits
Supplier:  VWR International
Certificates
VWR® Microdispenser Replacement Kits
VWR® Microdispenser Replacement Kits
5 Options Available Below
Product Details & Documents
The 5, 10, and 20 µl VWR® Microdispenser Replacement Kits each include 2 plungers, 2 O-rings, 1 collet, and 1 coil spring and barrel tube.
  • Available in a variety of sizes
 : The 20 and 25, 50, 100, 100/200, and 250 µl VWR® Microdispenser Replacement Kits each include 1 plunger, 2 O-rings, 1 collet, 3 fluoropolymer resin tips, and 1 coil spring and barrel tube.
Product Family Options
Información del producto
  • Availability
    Price
  • 53506-910
    10 uL VWR® Microdispenser Replacement Kit
    10 uL VWR® Microdispenser Replacement Kit
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (1UN)
    Each (1UN) - $952.34
    $952.34
    Each (1UN)
       $952.34
  • 53506-914
    20 and 25 uL VWR® Microdispenser Replacement Kit
    20 and 25 uL VWR® Microdispenser Replacement Kit
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (1UN)
    Each (1UN) - $952.34
    $952.34
    Each (1UN)
       $952.34
  • 53506-922
    50 uL VWR® Microdispenser Replacement Kit
    50 uL VWR® Microdispenser Replacement Kit
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (1UN)
    Each (1UN) - $936.72
    $936.72
    Each (1UN)
       $936.72
  • 53506-926
    100/200 uL VWR® Microdispenser Replacement Kit
    100/200 uL VWR® Microdispenser Replacement Kit
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (1UN)
    Each (1UN) - $952.34
    $952.34
    Each (1UN)
       $952.34
  • 53506-928
    2500 uL VWR® Microdispenser Replacement Kit
    2500 uL VWR® Microdispenser Replacement Kit
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (1UN)
    Each (1UN) - $952.34
    $952.34
    Each (1UN)
       $952.34