VWR® Glass Microanalysis Filter Holders
Filter Holder with Glass Support, 152 mm, 25 mm, 25 mm, 15 ml
Supplier: VWR International
Jump to:
Product Details & Documents
Used to filter under vacuum/small volumes for particulate or biological contamination analysis and for general filter application.
- Available in fritted glass or stainless steel
- Thick pre filters or laminated filters will not seal in this holder
Use durapore (PVDF) membrane or unlaminated PTFE for solvent applications with this apparatus.
Delivery information: The unit is supplied with a coarse porosity fritted or stainless-steel support base, 15 ml graduated funnel, anodized aluminum clamp, and a no.5 silicone stopper.
Documents
Certificates
Buscar por número de loteEspecificaciones
- Ø:25 mm
- Filter Diameter:25 mm
- Description:Filter Holder with Glass Support
- Height:152 mm
- Funnel Capacity:15 ml