Nunc® Externally-Threaded Universal Tubes, 1.8 ml, Thermo Scientific
Catalog # 89233-712
Supplier: Thermo Fisher Scientific
Specifications
- Volumen:1.8 ml
- Descripción:Sterile Cryo Tube with 2D Barcode
- Referencia:89233-712
- Empaquetado:Bulk Packaging
- Estilo:Flat Bottom
- Nº de proveedor:374502
Specifications
About this item
Versatile Nunc® 1.8 ml externally-threaded universal tubes can be used inside a large storage system, or in smaller scale applications such as traditional cryostorage in dewars.
All tubes and trays are supplied free from DNA, RNAse, DNAse, and endotoxins.
Certifications: IATA certification ensures that shipped samples will not leak.
Packaging: Available racked or in bulk, with 2D barcodes or non-coded.