ASTM Approved Non-Mercury 'Precision' Thermometers, Thermco
Proveedor: THERMCO PRODUCTS, INC.
Certificados
Acerca de este artículo
ASTM approved non-mercury blue spirit "precision" thermometers
Tenth of a scale division. The ice piont enables the user to validate and update the calibration document. Rages covered are from -38 to 155 degrees C and -36 to 311 degrees F.
Each incorporates an ice point on the scale, calibrated against NIST standards and certified at one temperature point.
Certifications: ASTM approved/ISO17025/ILAC-MRA accredited lab.
Ordering information: Accessories not included.
Especificaciones
- Catalog No:
- 10053-220
- 10053-224
- 10052-986
- 10053-454
- Environmentally Preferable:
- Safer Human and Environmental Health
- Safer Human and Environmental Health
- Safer Human and Environmental Health
- Safer Human and Environmental Health