Specifications
- Volumen:1.2 cu. ft.
- Color:Clear, with Gas Ports
- Descripción:2.0 Gas-Purge Desiccator Cabinet
- Referencia:47750-996
- La x An x Al:16.3×13.4×12.4"
- No. de bandejas:2
- Nº de proveedor:F42072-1002
Specifications
About this item
The attractive design and sturdy construction of the Secador® line of desiccator cabinets assures easy and reliable storage of moisture- and UV-sensitive products.
- Sturdy construction
- Easy and reliable storage
- Protects moisture- and UV-sensitive products
- Polycarbonate construction
Polycarbonate construction features a UV light blocker and is resistant to staining, crazing, and chemical attack.
Rear-mounted gas ports are tapered to accept tubing with ¼ to ⅜” I.D. Large doors maximize access to interior space and have a patented seal design, tab latches and security loops for a padlock or tamper-evident seals. Perforated shelves facilitate circulation of dry air and have space for desiccant under bottom shelf. Easy-to-view digital hygrometer is located in the front of the door that opens to the left.
Units have a space saving 34.1W x 41.4 cmD footprint and can be stacked up to 3 units high. U.S. Patents: 6,834,920; 7,318,630 and D474,626.