Qualitative Filter Paper, Grade 609, Ahlstrom
Catalog # 30617-436
Supplier: AHLSTROM MUNKSJO FILTRATION LLC.
Specifications
- Ø:33 cm
- Retencion:4.0 µm
- Capacidad de carga:Medium
- Velocidad de flujo:Medium
- Referencia:30617-436
- Nº de proveedor:6090-3300
Specifications
About this item
Ahlstrom Grade 609 is an evolution of Ahlstrom 613 for use in the beer industry.
This is a filter paper with an extremely low iron and nitrogen content for the analysis of protein in beer.