Ashless Quantitative Paper, Grade 74, Ahlstrom
Supplier: AHLSTROM MUNKSJO FILTRATION LLC.
Certificates
About this item
Ahlstrom Grade 74 is a medium flow rate ashless (Ash 0.007%) filter paper suitable for routine quantitative gravimetric techniques.
Specifications
- Catalog No:
- 47745-740
- 38006-106
- Velocidad de flujo:
- Medium
- Medium
- Capacidad de carga:
- Medium
- Medium
- Retencion:
- 2.0 µm
- 2.0 µm