pH Electrodes, IDS (Intelligent Digital Sensors), for Multi, WTW
97042-836
: YSI INC
- Rango de temperatura:0...+80 °C
- Descripción:IDS pH electrode Sentix® 940
- Matriz:Plastic
- Referencia:97042-836
- Rango de pH:0.000 - 14.000 (±0.004)
- Conector:1.5 m cable
- Nº de proveedor:103740
- Electrolyte:Gel
High quality, enhanced sensor technology combined with most modern measurement electronics.
- IDS electrodes include their serial number and their calibration records
- Measurement integrity benchmarking of the cells by QSC (Quality Sensor Control)
- Ø×L: 12×120 mm
: Various cable lengths available.