Orders LinkContactUsLinkComponent
Tote Box, Polypropylene, Dynalon
12×7 3/4×5 1/8" (L×W×H)
  470173-918
 :   DYNALON
 :  410535
Tote Box, Polypropylene, Dynalon
  470173-918
 :  DYNALON
Tote Box, Polypropylene, Dynalon
Tote Box, Polypropylene, Dynalon
12×7 3/4×5 1/8" (L×W×H)
$597.65
Each (1UN) Case de 6 (1UN)
Sterilizing Tote Boxes, molded of natural Polypropylene is suitable for use with food
These tote boxes are chemically resistant to acids, alkalies, oils and detergents and will not rust or corrode. Can be nested when stored.
  • Dimensions:
    12×7 3/4×5 1/8" (L×W×H)