Mini Benchtop Coolers
–12 °C Mini Cooler with gel filled lid, Blue, 245×105×100 mm, 32
Catalog # 89511-798
Supplier: HEATHROW SCIENTIFIC
Supplier #: HS120075
Jump to:
Product Details & Documents
Ideal for keeping samples, PCR reagents, enzymes, and other biochemical reagents cool while working at the bench.
- Protects temperature-sensitive samples at the bench or in the freezer
- Polycarbonate construction with rubber non-slip feet
- Gridded and keyed one-position lids for easy sample identification
- Temperature-retention graph included
Filled with insulating gel, the Mini Coolers can also be used for controlled freezing of samples with longer temperature retention.
Specifications
- Color:Blue
- Description:–12 °C Mini Cooler with gel filled lid
- Dimensions:245×105×100 mm
- No. of Places:32