DuraPorter™ Transport Box, Heathrow Scientific®
89511-802
- Color:Red with Red Handles
- Dimensiones:380×196×160 mm (15×7.7×6.3")
- Referencia:89511-802
- Nº de proveedor:HS120077
Ideal for transporting biological and clinical samples, as well as instruments and products that need to be kept clean and dry under testing environmental conditions
The clarity of the DuraPorter™ box enables users to visibly check sample integrity, so that it can be opened using the right precautions. All boxes feature leak-proof, polycarbonate construction. Lids open a full 180°.