Chemfluor® Fluoropolymer Tubing, PTFE, Saint-Gobain
Catalog # 63014-900
Supplier: SAINT GOBAIN PERFORMANCE PLASTICS
Specifications
- Tubo ext.Ø:⅛"
- Tubo Ø int.:¹⁄₁₆"
- Grosor de pared:0.031
- Modelo:63014-900
- Dureza en durómetro:58 (Shore D)
- Max. vacuum:29.9 in. Hg
- Working pressure:336.0 psi
- Referencia:63014-900
- Nº de proveedor:TSPTFE012531
Specifications
About this item
Chemfluor® tubings have outstanding chemical and physical characteristics because they are made using fluoropolymers and these materials are processed under stringently controlled conditions.
- Chemically inert
- Ultra-pure
- Non-toxic, non-stick
- Wide temperature capabilities
- Non-corroding and non-oxidizing
- Outstanding dielectric properties
- Autoclavable
In addition to being chemically inert, Chemfluor® tubings can be used in a broad range of temperatures, from −400 to +500 °F.
Ordering information: Length of package is 50 feet of translucent tubing.