ContactUsLinkComponent
MBP Electroporator Cuvettes, Molecular BioProducts
MBP Electroporator Cuvettes, Molecular BioProducts
Catálogo # 13500-364
undefined
MBP Electroporator Cuvettes, Molecular BioProducts
Catálogo # 13500-364
Número de proveedor:  5520
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Especificaciones
  • Descripción:
    Blue Cap
  • Referencia:
    13500-364
  • Gap Size:
    2 mm
  • Nº de proveedor:
    5520
Especificaciones
Acerca de este artículo
Molded from premium quality polycarbonate, our cuvettes are designed for maximum transformation efficiency
Snug-fitting cap offers easy handling and a color-coded, frosted surface for labeling. Pre-sterilized and individually packaged for added protection against contamination. Compatible with most common electroporation systems (Bio-Rad Gene Pulser, BTX, and others) and available with 1, 2, and 4 mm gap widths for all of your electroporation needs. Cuvettes are pre-sterilized and individually wrapped in packs of 50 each.