Orders LinkContactUsLinkComponent
MBP DNA AWAY™
MBP DNA AWAY™
DNA Away Squeeze Bottle, 250 mL
Catalog # 53509-506
Número de proveedor #:  7010
MBP DNA AWAY™
Catalog # 53509-506
CAS Number:  
MBP DNA AWAY™
MBP DNA AWAY™
DNA Away Squeeze Bottle, 250 mL
CAS Number:  
Price
$1,080.00
Log in Para disponibilidad
Each(1/Itm/Bx) Case de 12 (1/ Itm/Bx)
Product Details & Documents
RNase AWAY® eliminates RNase and DNA from laboratory surfaces
  • Easily Remove RNase Contaminants
  • Safe to Use on Pipettors, Glassware, Plasticware, and Most Metal Surfaces
  • Space-Saving, Convenient Packaging
  • Eliminates Over-Spraying of Decontaminants on Sensitive Equipment
The surface decontaminant is ideal for decontaminating apparatus, benchtops, glassware, and plasticware. The decontaminant leaves no residue to interfere with gel polymerization or staining and contains no strong acids. DNA AWAY™ decontaminant is ideal for removing DNase and DNA contamination.
Certifications: Biotechnology grade. Application Conformity Pass. Store frozen. Irritant. Hazard Statements: H315 / H319 / H335. Precautionary Statements: P280 / P302+P352 / P304+P340 / P305+P351+P338.
Especificaciones
Especificaciones
  • Description:
    DNA Away Squeeze Bottle
  • Size:
    250 mL